Please login to your online account to display your discounted pricing

anti-TNF-alpha Membrane Form, Fluorescein, Clone: 6401, R&D Systems


Manufacturer: r & d systems  FAB210F050

 View more versions of this product

Catalog No. FAB210F050

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha Membrane Form
Applications Flow Cytometry
Clone 6401
Conjugate Fluorescein
Cross Reactivity Human
Format Purified
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P01375
Host Species Murine
Immunogen &italic_begin;E. coli&italic_end;-derived recombinant human TNF-α
Isotype IgG1
Purification Method Protein A or G purified from ascites
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Test Specificity Detects cell surface expressed TNF-α (membrane and receptor bound forms) by flow cytometry. Detects human TNF-α in direct ELISAs and Western blots. In direct ELISAs, approximately 25‑50% cross‑reactivity with recombinant porcine TNF‑α and recombinant rhesus macaque TNF-alpha is observed but no cross-reactivity with recombinant cotton rat TNF‑alpha, recombinant rat TNF‑alpha, recombinant human (rh) LT alpha 1/beta 2, rhLT alpha 2/beta 1, rhAPRIL, rhBAFF, rhEDA‑A2, recombinant mouse EDA, rhFas Ligand, rhLIGHT, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, or rhVEGI is observed.
Target Species Human