Please login to your online account to display your discounted pricing

anti-Tmp21/p23, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 100-200). [Swiss-Prot No. P49755]
Antigen Tmp21/p23
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11057586T

 
novus biologicals canada
NB11057586SS
Unlabeled 0.025mL Each for $109.00

NB11057586

 
novus biologicals canada
NB11057586
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 100-200). [Swiss-Prot No. P49755]
Antigen Tmp21/p23
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human, primate, mouse, rat, cow, rabbit, and chicken embryo.
Gene Accession No. P49755
Gene ID (Entrez) 10972
Formulation PBS, 30% glycerol
Concentration 0.9mg/mL