Please login to your online account to display your discounted pricing

anti-TMEM97, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130436

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130436

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 27346
Concentration 1mg/mL
Gene Accession No. Q5BJF2
Quantity 0.1mL
Primary or Secondary Primary
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology.
Formulation PBS, 30% glycerol
Immunogen Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot No. Q5BJF2]
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Host Species Rabbit
Antigen TMEM97