Please login to your online account to display your discounted pricing

anti-TMEM97, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130437SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130437SS

  • / Each
 

Description & Specifications

Specifications

Antigen TMEM97
Applications Western Blot
Concentration 1.1mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q5BJF2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-Prot No. Q8VD00]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27346
Test Specificity Mouse. Immunogen sequence has 93% homology with rat.
Target Species Murine