Please login to your online account to display your discounted pricing

anti-TMEM97, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130437

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130437

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q5BJF2
Gene ID (Entrez) 27346
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Host Species Rabbit
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Mouse. Immunogen sequence has 93% homology with rat.
Concentration 1.1mg/mL
Cross Reactivity Mouse
Antigen TMEM97
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-Prot No. Q8VD00]
Quantity 0.1mL