missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TMED4, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing amino acids CNYRTQMWDKQKEV (amino aicds 55-67) of human HNLF. Genbank accession no. EAL23751.
Antigen TMED4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056393 Novus Biologicals Canada
NB10056393
0.1mg Each for $255.80
NB10056393T Novus Biologicals Canada
NB10056393SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TMED4
Purified
RUO
An approx. 26 kDa protein is recognized in HL60 cell lysate. Cross reacts with human. Predicted to cross react with mouse and rat.
222068
0.5mg/mL
A synthetic peptide containing amino acids CNYRTQMWDKQKEV (amino aicds 55-67) of human HNLF. Genbank accession no. EAL23751.
Polyclonal
Primary
Western Blot
Q7Z7H5
PBS containing 0.05% BSA