Please login to your online account to display your discounted pricing

anti-TLR9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP118284

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP118284

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology. Immunogen sequence has 92% homology to rat, 86% homology to sheep, porcine and bovine and 85% homology to mouse.
Antigen TLR9
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9NR96
Host Species Rabbit
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of the human TLR9 protein (within residues 1019-1032). [Swiss-Prot No. Q9NR96]
Gene ID (Entrez) 54106
Concentration 1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human