Please login to your online account to display your discounted pricing

anti-TLR9, Polyclonal, Novus Biologicals™

$111.78 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TLR9 protein (within residues 1019-1032). [Swiss-Prot No. Q9NR96]
Antigen TLR9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118284

 
novus biologicals canada
NBP118284
Unlabeled 0.1mL Each for $309.05

NBP118284SS

 
novus biologicals canada
NBP118284SS
Unlabeled 0.025mL Each for $111.78
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TLR9 protein (within residues 1019-1032). [Swiss-Prot No. Q9NR96]
Antigen TLR9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology. Immunogen sequence has 92% homology to rat, 86% homology to sheep, porcine and bovine and 85% homology to mouse.
Gene Accession No. Q9NR96
Gene ID (Entrez) 54106
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL