Please login to your online account to display your discounted pricing

anti-TLR7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107077SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107077SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51284
Concentration 1mg/mL
Test Specificity Human. Predicted to react with mouse, rat, feline, bovine, porcine, canine, and multiple other species based on 100% sequence homology.
Regulatory Status RUO
Gene Accession No. Q9NYK1
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of the human TLR7 protein (within residues 750-800). [Swiss-Prot No. Q9NYK1]
Antigen TLR7
Quantity 0.025mL
Host Species Rabbit
Format Affinity Purified
Cross Reactivity Human