Please login to your online account to display your discounted pricing

anti-TLR7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107077

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107077

  • / Each
 

Description & Specifications

Specifications

Antigen TLR7
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9NYK1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TLR7 protein (within residues 750-800). [Swiss-Prot No. Q9NYK1]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51284
Test Specificity Human. Predicted to react with mouse, rat, feline, bovine, porcine, canine, and multiple other species based on 100% sequence homology.
Target Species Human