Please login to your online account to display your discounted pricing

anti-TLR7, Polyclonal, Novus Biologicals™

$214.00 - $214.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 706-749 of human TLR7; GenBank no. gb|AAF78035.1|AF245702_1. It will cross-react with mouse TLR7. In human Ramos cells
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224905SS

 
novus biologicals canada
NBP224905SS
Unlabeled 0.025mg Each for $214.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 706-749 of human TLR7; GenBank no. gb|AAF78035.1|AF245702_1. It will cross-react with mouse TLR7. In human Ramos cells
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 51284
Formulation 100μg in 200μL PBS, contents are sterile
Concentration 0.5mg/mL