Please login to your online account to display your discounted pricing

anti-TLR7, Polyclonal, Novus Biologicals™

$111.78 - $321.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TLR7 protein (within residues 750-800). [Swiss-Prot No. Q9NYK1]
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP107077SS

 
novus biologicals canada
NBP107077SS
Unlabeled 0.025mL Each for $111.78

NBP107077

 
novus biologicals canada
NBP107077
Unlabeled 0.1mL Each for $321.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TLR7 protein (within residues 750-800). [Swiss-Prot No. Q9NYK1]
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human. Predicted to react with mouse, rat, feline, bovine, porcine, canine, and multiple other species based on 100% sequence homology.
Gene Accession No. Q9NYK1
Gene ID (Entrez) 51284
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL