Please login to your online account to display your discounted pricing

anti-TLR7, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 900-950 of human TLR7 was used as the immunogen.
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224767SS

 
novus biologicals canada
NBP224767SS
Unlabeled 0.025mg Each for $139.00

NBP224767

 
novus biologicals canada
NBP224767
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 900-950 of human TLR7 was used as the immunogen.
Antigen TLR7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 83% homologous in dog and 72% homologous in mouse.
Gene ID (Entrez) 51284
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL