Please login to your online account to display your discounted pricing

anti-TLR6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11093544SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11093544T

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Human, mouse, primate and rat. Immunogen sequence has 92% homology to sheep.
Format Affinity Purified
Applications Western Blotting
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR6 protein (within residues 700-796). [Swiss-Prot No. Q9Y2C9]
Antigen TLR6
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Gene ID (Entrez) 10333
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9Y2C9
Host Species Rabbit
Regulatory Status RUO
Quantity 0.025mL