Please login to your online account to display your discounted pricing

anti-TLR6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11093544

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11093544

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Gene ID (Entrez) 10333
Test Specificity Human, mouse, primate and rat. Immunogen sequence has 92% homology to sheep.
Formulation Tris-citrate/phosphate, pH 7-8
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Rabbit
Quantity 0.1mL
Gene Accession No. Q9Y2C9
Antigen TLR6
Regulatory Status RUO
Format Affinity Purified
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR6 protein (within residues 700-796). [Swiss-Prot No. Q9Y2C9]
Applications Western Blotting