Please login to your online account to display your discounted pricing

anti-TLR6, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR6 protein (within residues 700-796). [Swiss-Prot No. Q9Y2C9]
Antigen TLR6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11093544

 
novus biologicals canada
NB11093544
Unlabeled 0.1mL Each for $309.05

NB11093544T

 
novus biologicals canada
NB11093544SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR6 protein (within residues 700-796). [Swiss-Prot No. Q9Y2C9]
Antigen TLR6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, mouse, primate and rat. Immunogen sequence has 92% homology to sheep.
Gene Accession No. Q9Y2C9
Gene ID (Entrez) 10333
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL