Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056579SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056579T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen TLR4
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O00206
Host Species Rabbit
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A portion of amino acids 100-150 of human TLR4 was used as the immunogen.
Quantity 0.025mg
Gene ID (Entrez) 7099
Test Specificity Observed size: approx. 90 kD (native) or 80 kD (partial recombinant mouse TLR4--extracellular portion plus His-tag).
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO