Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056579

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056579

  • / Each
 

Description & Specifications

Specifications

Antigen TLR4
Formulation PBS containing 0.05% BSA
Gene Accession No. O00206
Host Species Rabbit
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7099
Test Specificity Observed size: approx. 90 kD (native) or 80 kD (partial recombinant mouse TLR4--extracellular portion plus His-tag).
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Immunogen A portion of amino acids 100-150 of human TLR4 was used as the immunogen.
Regulatory Status RUO