Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224537

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224537

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. O00206
Applications Western Blotting
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7099
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, rat, mouse, dog, chicken and chimpanzee, and 94% homologous in zebrafish and elephant shark.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Antigen TLR4
Format Affinity Purified
Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human TLR4 was used as the immunogen for this antibody.
Regulatory Status RUO