Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178427SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178427SS

  • / Each
 

Description & Specifications

Specifications

Antigen TLR4
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A synthetic peptide made to the C-terminal portion of the human TLR4 protein (between residues 650-710) [UniProt O00206]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7099
Concentration 1.14mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat