Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178427

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178427

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene ID (Entrez) 7099
Immunogen A synthetic peptide made to the C-terminal portion of the human TLR4 protein (between residues 650-710) [UniProt O00206]
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Concentration 1.14mg/mL
Quantity 0.1mL
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit
Antigen TLR4