Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human TLR4 was used as the immunogen for this antibody.
Antigen TLR4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224538

 
novus biologicals canada
NBP224538
Unlabeled 0.1mg Each for $307.00

NBP224538SS

 
novus biologicals canada
NBP224538SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human TLR4 was used as the immunogen for this antibody.
Antigen TLR4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Reacts with Bison, Chimpanzee, Cow, Gorilla, Horse, Human, Macaque. Predicted reactivity with Rat, Dog.
Gene Accession No. O00206
Gene ID (Entrez) 7099
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL