Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human TLR4 was used as the immunogen for this antibody.
Antigen TLR4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224537SS

 
novus biologicals canada
NBP224537SS
Unlabeled 0.025mg Each for $109.00

NBP224537

 
novus biologicals canada
NBP224537
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human TLR4 was used as the immunogen for this antibody.
Antigen TLR4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, rat, mouse, dog, chicken and chimpanzee, and 94% homologous in zebrafish and elephant shark.
Gene Accession No. O00206
Gene ID (Entrez) 7099
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL