missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TLR4, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human TLR4 was used as the immunogen.
Antigen TLR4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056579T Novus Biologicals Canada
NB10056579SS
0.025mg Each for $139.00
NB10056579 Novus Biologicals Canada
NB10056579
0.1mg Each for $307.00
Specifications Description & Specifications

Specifications

Rabbit
TLR4
Purified
RUO
Observed size: approx. 90 kD (native) or 80 kD (partial recombinant mouse TLR4--extracellular portion plus His-tag).
7099
0.5mg/mL
A portion of amino acids 100-150 of human TLR4 was used as the immunogen.
Polyclonal
Primary
Immunoprecipitation,Western Blot
O00206
PBS containing 0.05% BSA