Please login to your online account to display your discounted pricing

anti-TLR4 Interactor With Leucine-Rich Repeats, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227168SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227168SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Gene Accession No. Q7L0X0
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9865
Antigen TLR4 Interactor With Leucine-Rich Repeats
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is homologous in human (100%), mouse (92%) and rat (92%) TRIL.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 770-820 of human TRIL was used as the immunogen for this antibody.
Quantity 0.025mg
Cross Reactivity Human