Please login to your online account to display your discounted pricing

anti-TLR4 Interactor With Leucine-Rich Repeats, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227168

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227168

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is homologous in human (100%), mouse (92%) and rat (92%) TRIL.
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9865
Concentration 0.5mg/mL
Format Purified
Immunogen A portion of amino acids 770-820 of human TRIL was used as the immunogen for this antibody.
Applications Western Blotting
Quantity 0.1mg
Host Species Rabbit
Gene Accession No. Q7L0X0
Primary or Secondary Primary
Antigen TLR4 Interactor With Leucine-Rich Repeats
Regulatory Status RUO
Cross Reactivity Human