missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TLR4 Interactor With Leucine-Rich Repeats, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 770-820 of human TRIL was used as the immunogen for this antibody.
Antigen TLR4 Interactor With Leucine-Rich Repeats
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227168 Novus Biologicals Canada
NBP227168
0.1mg Each for $257.41
NBP227168SS Novus Biologicals Canada
NBP227168SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
TLR4 Interactor With Leucine-Rich Repeats
Purified
RUO
The amino acid sequence used as immunogen is homologous in human (100%), mouse (92%) and rat (92%) TRIL.
9865
0.5mg/mL
A portion of amino acids 770-820 of human TRIL was used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
Q7L0X0
100μg antibody in 200μL PBS containing 0.05% BSA.