Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224565

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224565

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, opossum, mouse, rat, horse, pig, dog, chicken and chimpanzee.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Antigen TLR3
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A peptide containing H-539 and N-541
Gene ID (Entrez) 7098
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit