Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11093591SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11093591T

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Rabbit
Applications Western Blotting
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR3 protein (within residues 800-904). [Swiss-Prot No. O15455]
Primary or Secondary Primary
Concentration 1mg/mL
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. O15455
Quantity 0.025mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7098
Test Specificity Human, primate, porcine and feline. 92% sequence identity with mouse, rat, equine and bovine.
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Antigen TLR3