Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227167SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227167SS

  • / Each
 

Description & Specifications

Specifications

Antigen TLR3
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen The immunogen for this antibody was a portion of amino acids 325-393 of mouse TLR3. 356 373 of mouse TLR3
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7098
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 83% in guinea pig, 72% in bovine, horse and rat, 67% in rabbit and human, and 61% homologous in pig.
Target Species Human