Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen The immunogen for this antibody was a portion of amino acids 325-393 of mouse TLR3. 356 373 of mouse TLR3
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227167

 
novus biologicals canada
NBP227167
Unlabeled 0.1mg Each for $259.02

NBP227167SS

 
novus biologicals canada
NBP227167SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The immunogen for this antibody was a portion of amino acids 325-393 of mouse TLR3. 356 373 of mouse TLR3
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 83% in guinea pig, 72% in bovine, horse and rat, 67% in rabbit and human, and 61% homologous in pig.
Gene ID (Entrez) 7098
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL