Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™

$142.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A peptide containing H-539 and N-541
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224565

 
novus biologicals canada
NBP224565
Unlabeled 0.1mg Each for $307.00

NBP224565SS

 
novus biologicals canada
NBP224565SS
Unlabeled 0.025mg Each for $142.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide containing H-539 and N-541
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, opossum, mouse, rat, horse, pig, dog, chicken and chimpanzee.
Gene ID (Entrez) 7098
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL