Please login to your online account to display your discounted pricing

anti-TLR3, Polyclonal, Novus Biologicals™

$106.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR3 protein (within residues 800-904). [Swiss-Prot No. O15455]
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11093591T

 
novus biologicals canada
NB11093591SS
Unlabeled 0.025mL Each for $106.00

NB11093591

 
novus biologicals canada
NB11093591
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human TLR3 protein (within residues 800-904). [Swiss-Prot No. O15455]
Antigen TLR3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, primate, porcine and feline. 92% sequence identity with mouse, rat, equine and bovine.
Gene Accession No. O15455
Gene ID (Entrez) 7098
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL