Please login to your online account to display your discounted pricing

anti-TLR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056586SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056586T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_003255
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Gene ID (Entrez) 7097
Conjugate Unlabeled
Immunogen This antibody was developed against a portion of the extracellular region of mouse TLR2.
Quantity 0.025mg
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Concentration 0.5mg/mL
Antigen TLR2
Format Purified
Host Species Rabbit
Cross Reactivity Mouse