Please login to your online account to display your discounted pricing

anti-TLR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056586

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056586

  • / Each
 

Description & Specifications

Specifications

Antigen TLR2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_003255
Host Species Rabbit
Immunogen This antibody was developed against a portion of the extracellular region of mouse TLR2.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7097
Target Species Murine