Please login to your online account to display your discounted pricing

anti-TLR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056573SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056573T

  • / Each
 

Description & Specifications

Specifications

Antigen TLR2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. NP_003255
Host Species Rabbit
Immunogen This antibody was developed against an extracellular domain of mouse TLR2 (amino acids 564-580 and 751-770).
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7097
Test Specificity An 86 kDa band is observed.
Target Species Human
Target Species Murine