Please login to your online account to display your discounted pricing

anti-TLR2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056573

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056573

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7097
Concentration 0.5mg/mL
Test Specificity An 86 kDa band is observed.
Gene Accession No. NP_003255
Antigen TLR2
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Conjugate Unlabeled
Quantity 0.1mg
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen This antibody was developed against an extracellular domain of mouse TLR2 (amino acids 564-580 and 751-770).
Regulatory Status RUO
Applications Western Blotting
Host Species Rabbit