Please login to your online account to display your discounted pricing

anti-TLR15, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227191

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227191

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL; crude rabbit sera with 50% glycerol.
Gene Accession No. Q2XQ10
Antigen TLR15
Host Species Rabbit
Immunogen Amino acids 164-190 of chicken TLR15 protein were used as the immunogen.
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 421219
Test Specificity The amino acid sequence used as immunogen is 100% homologous in chicken.
Conjugate Unlabeled
Format Antiserum
Applications Peptide ELISA
Regulatory Status RUO
Cross Reactivity Chicken