Please login to your online account to display your discounted pricing

anti-TLR15, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Amino acids 164-190 of chicken TLR15 protein were used as the immunogen.
Antigen TLR15
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227191SS

 
novus biologicals canada
NBP227191SS
Unlabeled 0.025mL Each for $109.00

NBP227191

 
novus biologicals canada
NBP227191
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 164-190 of chicken TLR15 protein were used as the immunogen.
Antigen TLR15
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Peptide ELISA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in chicken.
Gene Accession No. Q2XQ10
Gene ID (Entrez) 421219
Formulation 100μL; crude rabbit sera with 50% glycerol.