Please login to your online account to display your discounted pricing

anti-TLR15, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227191SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227191SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Format Antiserum
Formulation 100μL; crude rabbit sera with 50% glycerol.
Gene Accession No. Q2XQ10
Host Species Rabbit
Immunogen Amino acids 164-190 of chicken TLR15 protein were used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 421219
Test Specificity The amino acid sequence used as immunogen is 100% homologous in chicken.
Cross Reactivity Chicken
Antigen TLR15
Applications Peptide ELISA