Please login to your online account to display your discounted pricing

anti-TLR12, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids of 743-756 (RVWLQGLRGKGDKG) of mouse TLR12.
Antigen TLR12
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224833SS

 
novus biologicals canada
NBP224833SS
Unlabeled 0.025mg Each for $139.00

NBP224833

 
novus biologicals canada
NBP224833
Unlabeled 0.1mg Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids of 743-756 (RVWLQGLRGKGDKG) of mouse TLR12.
Antigen TLR12
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Gene ID (Entrez) 100131451
Formulation 100μg in 200μLs PBS containing 0.2% gelatin
Concentration 0.5mg/mL