Please login to your online account to display your discounted pricing

anti-TLR11, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056742SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056742T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Antigen TLR11
Format Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 442887
Test Specificity Immunizing peptide will block the mouse TLR11 band in western blotting. The approx. 32 kDa band may be a proteolytic degradation product of TLR11.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Rabbit
Applications Flow Cytometry
Applications Western Blotting
Immunogen A synthetic peptide within amino acid residues 911-926 (CEQLKRRLSKAGQERD) of mouse TLR11 (Genbank accession no. NP_991388) was used as immunogen.
Quantity 0.025mg