Please login to your online account to display your discounted pricing

anti-TLR11, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056742

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056742

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Test Specificity Immunizing peptide will block the mouse TLR11 band in western blotting. The approx. 32 kDa band may be a proteolytic degradation product of TLR11.
Format Purified
Applications Flow Cytometry
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 442887
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen TLR11
Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 911-926 (CEQLKRRLSKAGQERD) of mouse TLR11 (Genbank accession no. NP_991388) was used as immunogen.
Quantity 0.1mg