Please login to your online account to display your discounted pricing

anti-TLR10, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10080840

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10080840

  • / Each
 

Description & Specifications

Specifications

Antigen TLR10
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS
Gene Accession No. Q9BXR5
Host Species Rabbit
Immunogen Synthetic peptide; peptide corresponding to 14 amino acids near the middle of human TLR10
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 81793
Target Species Human