Please login to your online account to display your discounted pricing

anti-TLR10, Polyclonal, Novus Biologicals™

$371.80 - $371.80

Specifications

Host Species Rabbit
Immunogen Synthetic peptide; peptide corresponding to 14 amino acids near the middle of human TLR10
Antigen TLR10
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10080840

 
novus biologicals canada
NB10080840
Unlabeled 0.05mg Each for $371.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide; peptide corresponding to 14 amino acids near the middle of human TLR10
Antigen TLR10
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q9BXR5
Gene ID (Entrez) 81793
Formulation PBS
Concentration 1mg/mL