Please login to your online account to display your discounted pricing

anti-TLR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056563SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056563T

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene ID (Entrez) 7096
Gene Accession No. NP_003254
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Format Purified
Quantity 0.025mg
Test Specificity In Ramos, an 87 kDa band is observed.
Antigen TLR1
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide corresponding to a sequence within amino acids 400-450 of human TLR1.