Please login to your online account to display your discounted pricing

anti-TLR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056563

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056563

  • / Each
 

Description & Specifications

Specifications

Antigen TLR1
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. NP_003254
Host Species Rabbit
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide corresponding to a sequence within amino acids 400-450 of human TLR1.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7096
Test Specificity In Ramos, an 87 kDa band is observed.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat