Please login to your online account to display your discounted pricing

anti-TIP60, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 480-530 of human TIP60 was used as the immunogen.
Antigen TIP60
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224613

 
novus biologicals canada
NBP224613
Unlabeled 0.1mg Each for $259.02

NBP224613SS

 
novus biologicals canada
NBP224613SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 480-530 of human TIP60 was used as the immunogen.
Antigen TIP60
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b, CRA_c, CRA_d and CRA_e) and chimpanzee, chicken, dog, mouse and rat, 95% homologous in monkey and xenopus, 85% homologous in zebrafish and 80% homologous in pufferfish.
Gene Accession No. Q92993
Gene ID (Entrez) 10524
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL