missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TIM-3, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NB10056422SS

 View more versions of this product

Catalog No. NB10056422T


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

TIM-3
0.5mg/mL
PBS containing 0.05% BSA
Rabbit
0.025mg
Primary
84868
Murine
Western Blot
Unlabeled
Purified
A portion of amino acids 100-200 of mouse TIM-3 protein were used as the immunogen.
RUO
Polyclonal
The NCBI accession number for TIM-3 is AAL35776. Reacts with Mouse. Predicted to react with Rat (79%).