Please login to your online account to display your discounted pricing

anti-TIM-3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056422

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056422

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84868
Test Specificity The NCBI accession number for TIM-3 is AAL35776. Reacts with Mouse. Predicted to react with Rat (79%).
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Antigen TIM-3
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Format Purified
Applications Western Blotting
Immunogen A portion of amino acids 100-200 of mouse TIM-3 protein were used as the immunogen.